Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPIV/CD26 Mouse anti-Human, PE/Cy5.5, Clone: OTI11D7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270588PECY55

Catalog No. N27058PECY5


Add to cart

Description

Description

DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

DPPIV/CD26
OTI11D7
Flow Cytometry
Affinity Purified, Conjugated
DPP4
Full length recombinant protein of human DPPIV/CD26 (NP_001926) produced in HEK293T cells.
Immunogen affinity purified
Adaptive Immunity, Cellular Markers, GPCR, Immunology
Primary
1803.0
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5
Mouse
IgG1
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit