Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPS Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15464620UL

 View more versions of this product

Catalog No. NBP15464620


Add to cart

Description

Description

DPS Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

DPS
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
PDSS1
Synthetic peptides corresponding to PDSS1(prenyl (decaprenyl) diphosphate synthase, subunit 1) The peptide sequence was selected from the middle region of PDSS1. Peptide sequence VIDDASSRRGKHTVNKIWGEKKAVLAGDLILSAASIALARIGNTTVISIL.
46 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This product is specific to Subunit or Isoform: 1.
Western Blot
Western Blot 1:100-1:2000
Q5T2R2
All-trans-decaprenyl-diphosphate synthase subunit 1, COQ1SPS, Decaprenyl pyrophosphate synthase subunit 1, decaprenyl-diphosphate synthase subunit 1, DPS, DPS1, EC 2.5.1.91, hDPS1, MGC70953, polyprenyl pyrophosphate synthetase, prenyl (decaprenyl) diphosphate synthase, subunit 1, RP13-16H11.3, subunit 1 of decaprenyl diphosphate synthase, TPRT, TPT 1, TPTcoenzyme Q1 homolog, trans-prenyltransferase, trans-prenyltransferase (TPT), Trans-prenyltransferase 1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
23590
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit