Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPYD Antibody (SR1055), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32232950UL
Description
DPYD Monoclonal antibody specifically detects DPYD in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
DPYD | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1055 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
DHP, DHPDHase, dihydropyrimidine dehydrogenase, dihydropyrimidine dehydrogenase [NADP+], Dihydrothymine dehydrogenase, Dihydrouracil dehydrogenase, DPDMGC132008, EC 1.3.1.2, MGC70799 | |
A synthesized peptide derived from human DPYD (Uniprot #: Q12882) | |
50 μg | |
Amino Acids Drugs and other small molecules, Endocrinology, Signal Transduction | |
1806 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction