Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPYS Monoclonal Antibody (3B1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518759
Description
Peptide Sequence: AVNFNIFEGM VCHGVPLVTI SRGKVVYEAG VFSVTAGDGK FIPRKPFAEY IYKRIKQRDR TCTPTPVERA PYKGEVATLK SRVTKEDATA GTRKQAHP.
Dihydropyrimidinase catalyzes the conversion of 5,6-dihydrouracil to 3-ureidopropionate in pyrimidine metabolism. Dihydropyrimidinase is expressed at a high level in liver and kidney as a major 2.5-kb transcript and a minor 3.8-kb transcript. Defects in the DPYS gene are linked to dihydropyrimidinuria.
Specifications
DPYS | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q14117 | |
Dpys | |
DPYS (NP_001376, 422 a.a. ~ 519 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
3B1 | |
Unconjugated | |
Dpys | |
1200017I10Rik; 1300004I01Rik; DHP; DHPase; dihydropyrimidinase; Dihydropyrimidine amidohydrolase; Dpys; Hydantoinase | |
Mouse | |
Affinity Chromatography | |
RUO | |
1807 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction