Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPYS Mouse anti-Human, Clone: 8D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191291

 View more versions of this product

Catalog No. NBP191291


Add to cart

Description

Description

DPYS Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

DPYS
8D10
Western Blot 0.5 ug/ml
NP_001376
DHPasehydantoinase, DHPHydantoinase, dihydropyrimidinase, Dihydropyrimidine amidohydrolase, EC 3.5.2.2
Mouse
56 kDa
100ul
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution. Final concentration is 1mg/mL in PBS buffer.
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
DPYS
The immunogen for this antibody is DPYS.
Protein A or G purified
RUO
Primary
1807
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit