Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPYS Mouse anti-Human, Clone: 8D12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19129220UL

Catalog No. NBP1929220


Add to cart

Description

Description

DPYS Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

DPYS
8D12
Western Blot 1.25 ug/ml
NP_001376
DHPasehydantoinase, DHPHydantoinase, dihydropyrimidinase, Dihydropyrimidine amidohydrolase, EC 3.5.2.2
Mouse
IgG1 Kappa
Protein A or G purified
RUO
Primary
1807
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
DPYS
The immunogen for this antibody is DPYS.
57 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit