Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPYS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548576

Catalog No. PA548576

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Dihydropyrimidinase catalyzes the conversion of 5,6-dihydrouracil to 3-ureidopropionate in pyrimidine metabolism. Dihydropyrimidinase is expressed at a high level in liver and kidney as a major 2.5-kb transcript and a minor 3.8-kb transcript. Defects in the DPYS gene are linked to dihydropyrimidinuria.
Specifications

Specifications

DPYS
Polyclonal
PBS with 0.09% sodium azide
Q14117
Dihydropyrimidinase, DHP, DHPase, Dihydropyrimidine amidohydrolase, Hydantoinase, DPYS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1807
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DPYS
Liquid
DPYS
KLH conjugated synthetic peptide between 454-482 amino acids from the C-terminal region of human DPYS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit