Learn More
DPYSL2 Mouse anti-Human, Mouse, Clone: 33CT11.40.3, Abnova™
Mouse monoclonal antibody raised against human DPYSL2.
Manufacturer:  Abnova Corporation MAB12580
Catalog No. 89-934-052
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
DPYSL2
33CT11.40.3
Mouse monoclonal antibody raised against human DPYSL2.
In ascites (0.09% sodium azide).
CRMP2/DHPRP2/DRP-2/DRP2
Mouse
IgG2b
RUO
Primary
1808
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
DPYSL2
A synthetic peptide corresponding to amino acids 463-507 of human DPYSL2.
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine