DR 2313, Tocris

PARP-1 and PARP-2 inhibitor

Manufacturer: R&D Systems 2496/10

Quantity 10mg

 View more versions of this product

Catalog No. 24-961-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C8H10N2OS
    Formula Weight 182.24
    Synonym 1,5,7,8-Tetrahydro-2-methyl-4H-thiopyrano[4,3-d]pyrimidin-4-one
    Chemical Name or Material DR 2313

    Specifications

    Formula Weight 182.24
    Quantity 10mg
    Synonym 1,5,7,8-Tetrahydro-2-methyl-4H-thiopyrano[4,3-d]pyrimidin-4-one
    Molecular Formula C8H10N2OS
    CAS 284028-90-6
    Assay Percent Range >99%
    Recommended Storage Store at +4°C
    Chemical Name or Material DR 2313

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit