Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR 2313, Tocris Bioscience™

Click to view available options
Quantity:
1mg
10mg
50mg
This item is not returnable. View return policy
284028-90-6
C8H10N2OS
182.241
HRYKZAKEAVZGJD-UHFFFAOYSA-N
7,8-dihydro-2-methyl-1h-thiopyrano 4,3-d pyrimidin-4 5h-one,parp inhibitor xi, dr2313,1,5,7,8-tetrahydro-2-methyl-4h-thiopyrano 4,3-d pyrimidin-4-one,2-methyl-3,5,7,8-tetrahydro-4h-thiopyrano 4,3-d pyrimidin-4-one,2-methyl-7,8-dihydro-1h-thiopyrano 4,3-d pyrimidin-4 5h-one,3,5,7,8-tetrahydro-2-methyl-4h-thiopyrano 4,3-d pyrimidin-4-one,2-methyl-3,5,7,8-tetrahydrothiopyrano 4,3-d pyrimidine-4-one,2-methyl-1h,5h,7h,8h-thiopyrano 4,3-d pyrimidin-4-one,4pnl,drl
10219702
2-methyl-1,5,7,8-tetrahydrothiopyrano[4,3-d]pyrimidin-4-one
CC1=NC(=O)C2=C(N1)CCSC2
This item is not returnable. View return policy
284028-90-6
C8H10N2OS
182.241
HRYKZAKEAVZGJD-UHFFFAOYSA-N
7,8-dihydro-2-methyl-1h-thiopyrano 4,3-d pyrimidin-4 5h-one,parp inhibitor xi, dr2313,1,5,7,8-tetrahydro-2-methyl-4h-thiopyrano 4,3-d pyrimidin-4-one,2-methyl-3,5,7,8-tetrahydro-4h-thiopyrano 4,3-d pyrimidin-4-one,2-methyl-7,8-dihydro-1h-thiopyrano 4,3-d pyrimidin-4 5h-one,3,5,7,8-tetrahydro-2-methyl-4h-thiopyrano 4,3-d pyrimidin-4-one,2-methyl-3,5,7,8-tetrahydrothiopyrano 4,3-d pyrimidine-4-one,2-methyl-1h,5h,7h,8h-thiopyrano 4,3-d pyrimidin-4-one,4pnl,drl
10219702
2-methyl-1,5,7,8-tetrahydrothiopyrano[4,3-d]pyrimidin-4-one
CC1=NC(=O)C2=C(N1)CCSC2

PARP-1 and PARP-2 inhibitor

Competitive inhibitor of poly(ADP-ribose) polymerase (PARP) (IC 50 values are 0.20 and 0.24 μM for PARP-1 and PARP-2 respectively). Neuroprotective; reduces neuronal cell death in models of cerebral ischemia in vivo and in vitro. Brain penetrant.
Quantity 1mg
Formula Weight 182.24
Percent Purity >99%
Chemical Name or Material DR 2313

Recommended Storage : Store at +4°C