Learn More
Tocris Bioscience™ DR 4485 Hydrochloride
High affinity and selective 5-HT7 antagonist; orally bioavailable
$285.36 - $1194.80
Specifications
Chemical Name or Material | 6-Chloro-2a-[4-[4-(4-chlorophenyl)-3,6-dihydro-1(2H)-pyridinyl]butyl]-2a,3,4,5-tetrahydrobenz[cd]indol-2(1H)-one hydrochloride |
---|---|
CAS | 402942-53-4 |
Molecular Formula | C26H28Cl2N2O.HCl |
Formula Weight | 491.88 |
Purity | 98% |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-051-0
|
Tocris Bioscience™
5005/10 |
10 mg |
Each for $285.36
|
|
|||||
50-055-0RD
|
Tocris Bioscience™
5005/50 |
50 mg |
Each for $1,194.80
|
|
|||||
Description
- High affinity and selective 5-HT7 antagonist (pKi = 8.14)
- Exhibits selectivity for 5-HT7 over other 5-HT receptors
- Inhibits 5-HT-induced cAMP accumulation in HEK-293 cells expressing the 5-HT7 receptor
- Orally bioavailable
Specifications
6-Chloro-2a-[4-[4-(4-chlorophenyl)-3,6-dihydro-1(2H)-pyridinyl]butyl]-2a,3,4,5-tetrahydrobenz[cd]indol-2(1H)-one hydrochloride | |
C26H28Cl2N2O.HCl | |
98% |
402942-53-4 | |
491.88 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.