Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dr. FuelCell Model Cars
Teach concepts such as electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps.
Specifications
For Use With (Application) | Teach electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps |
---|---|
Grade | 8 to 12 |
Storage Requirements | 10° to 35°C |
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Includes | Price | Quantity & Availability | ||||
S66135
![]() |
Heliocentris
K000354EN |
Complete Kit | Car chassis with electric motor, fuel cell, solar panel, car cassis, storage case, cables, and lesson book, load measurement box |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Water and sunlight power a simple and effective demonstration of solar hydrogen technology
- Reversible PEM fuel cell works as both an electrolyzer and a fuel cell
- As an electrolyzer, it uses power from a solar module to split water into hydrogen and oxygen
- As a fuel cell, it uses stored hydrogen and oxygen to power the electric motor and drive the car
- Load measurement box has a voltmeter, ammeter, two electrical loads (lamp and electric motor) and seven different adjustable resistances for characteristic curve analysis
- Course book is written by an award-winning teacher; contains eight experiments designed with teacher and student in mind
- Grades 8-12
Specifications
Teach electrolysis, solar power, fuel cells, energy and energy conversion, volts, watts and amps | |
8 to 12 | |
10° to 35°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title