Learn More
Dräger™ Aerotest Multitest Kit
Used for detecting contaminants in compressed gases
Manufacturer:  Dräger™ 4056181
Catalog No. 19-046-638
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Multitest ≤75psi, complete
  • Reduces pressure of gas sample to level at which Draeger-Tubes™ can accurately operate
  • Includes: deluxe carrying case, instruction booklet, measuring device for 7 Tubes, quick-connect coupling, tube opener, timer and Draeger-Tubes™
Specifications
Specifications
Safety
Aerotest multitest kit