Learn More
Dräger™ CMS Chips: Formaldehyde
Suitable for 10 measurements of one gas
Supplier: Dräger™ 6406540
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
2 to 12mg/L | |
100 to 600 sec. | |
10°C to 30°C |
Formaldehyde CMS Chip | |
0.2 to 5 | |
50°F to 86°F |