Dräger™ Diffusion Tubes with Direct Indication
Convenient, long-term TWA monitoring—direct reading, with no lab analysis required
Manufacturer: Dräger™ 6733111
 View more versions of this product
Catalog No. 17-985-353K
  Check Availability
Add to Cart
    Specifications
    Humidity Range 3mg/L
    Measurement Range 1.3 to 25
    Measurement Time 8 hr.
    Quantity 10
    Temperature (English) Range 64.4° to 71.6°F
    Temperature (Metric) Range 18° to 22°C
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback