Dräger™ Individual Haz Mat Simultaneous Test Sets
For simultaneous testing of five different gases
Manufacturer: Dräger™ 8101735
 View more versions of this product
Catalog No. 18-556R
  Check Availability
Add to Cart
Alert
No other type of tube should be substituted with these kits.
Required Accessories : Require adapter and a Dräger pump (sold separately) for the semi-quantitative determination of inorganic gases and organic vapors.
    Specifications
    Includes Tests for hydrogen chloride, hydrogen cyanide, carbon monoxide, ammonia, and nitrous fumes
    Product Type Simultaneous test set I
    For Use With (Application) For simultaneous testing of five different gases
  • Description & Specifications
Related Products
Product Content Feedback