Dräger™ Installed XXS Dräger™Sensor™ Sensors for X-am™ 5000 Multigas Monitors
Manufacturer: Dräger™ 4543767
 View more versions of this product
Catalog No. 17-985-638
  Check Availability
Add to Cart
    Specifications
    Warranty 1 Year
    Measurement Range 0 to 50
    Resolution 1
    Sensor Types Hydrogen Cyanide (XXS)
    Temperature (English) Range -4° to +122°F
    Temperature (Metric) Range -20° to +50°C
  • Description & Specifications
Related Products
Product Content Feedback