Learn More
Dräger™ Multiwarn™ II Deluxe Multigas Monitor Preconfigured Kits
Supplier: Dräger™ 4523043
Description
Multiwarn II Monitors are also available in preconfigured kits which include sensors, calibration adapter, pump adapter switch, 12 in. (30.5cm) probe with water and dust filter, and a 3 ft. (0.9m) sample hose.
- Basic kit includes sensors for CO and H2S
- Standard kit comes with datalogger and IR sensor for combustible gases or CO2