Learn More
Dräger™ Multiwarn™ II Deluxe Multigas Monitor Preconfigured Kits
Manufacturer:  Dräger™ 4523057X
 View more versions of this product
Catalog No. 19-452-057
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Multiwarn II Monitors are also available in preconfigured kits which include sensors, calibration adapter, pump adapter switch, 12 in. (30.5cm) probe with water and dust filter, and a 3 ft. (0.9m) sample hose.
  • Basic kit includes sensors for CO and H2S
  • Standard kit comes with datalogger and IR sensor for combustible gases or CO2
Specifications
Specifications
Safety
Documents
Accessories