Learn More
Dräger™ Multiwarn™ II Deluxe Multigas Monitors
Supplier: Dräger™ 4523002
Includes: All monitors include internal pump and a NiCd battery pack with an eight-hour runtime per charge
Description
Basic model does not include sensors. To build a complete system, select a monitor from the Multiwarn II Monitors and add the desired “installed« sensors (sold separately).