Learn More
Dräger Multiwarn II Deluxe Multigas Monitors
Includes: All monitors include internal pump and a NiCd battery pack with an eight-hour runtime per charge
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
19-452001
|
Dräger
4523001 |
N/A | Basic monitor; w/o datalogger |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
19-452002
|
Dräger
4523002 |
N/A | Basic monitor; w/datalogger |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Basic model does not include sensors. To build a complete system, select a monitor from the Multiwarn II Monitors and add the desired “installed' sensors (sold separately).