Dräger™ NIOSH Respirator Cartridges

Manufacturer: Dräger™ 6737013

 View more versions of this product

Catalog No. 19-059-136

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Quantity 6 Pack
    For Use With (Equipment) X-plore™ 6530,X-plore™ 6530
    Product Type Respirator Filter
    For Use With (Application) To protect against all particulate aerosols, even those with oils present
    Certifications NIOSH
    Connection Type 40mm round bottle thread
    Filter Type P100 Filter
    Material (Housing) Aluminum
    Color Pink

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit