Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dräger™ Pac™ 3500 Single-Gas Detector
Offers two-year, maintenance-free use
Supplier: Dräger™ 4543959
Includes: Alligator clip for secure attachment
Description
Ideal for personal monitoring of industrial oxygen, carbon monoxide, or hydrogen sulfide.
- Compact, robust and ergonomic design with intuitive two-button operation
- Hardy sensors can be used in extreme environmental conditions without triggering false alarms
- Large, language-free backlit display continuously indicates gas concentration while simultaneously displaying the corresponding measurement unit
- Equipped with a sensor that allows gas intake from both the top and front of the unit to ensure continuous measurement, even if a gas inlet is covered
- Triple alarms for exceptional safety—clear, multi-toned signal, a bright, flashing 360° LED on top and bottom, and vibration
- Store up to 60 events, along with dates and times; infrared interface enables data to be loaded onto a PC for analysis (requires connecting cradle or E-Cal System and software, sold separately)
- When bump mode is activated, the unit automatically reminds users when a bump test or calibration is required
- Operates in a temperature range from -22° to +122°F (-30° to +50°C); relative humidity, 10 to 90%
Order Info
Dimensions: 0.8L x 2.5W x 3.3 in.H (2.0 x 6.4 x 8.4cm). Weight: 3.8 oz. (106g).
Certifications
IP65 rated for ingresss protection. UL Classified as Intrinsically Safe for use in Class I, Division 1, Group A, B, C, D, E, F, G hazardous locations, Temp Code T4; ATEX, I/II 1/2 G EEx ia I/IIC, T4. CE mark.
Compliance
Complies with EMC Directive 89/336/EEC.
Specifications
Audible, visual, vibrating | |
1 in. | |
Oxygen | |
6.4 cm | |
Alligator clip for secure attachment | |
0 to 25% | |
0.1% | |
-20°F to +120°F | |
3.8 oz. | |
3.3 in. |
8 hr. | |
2.5 cm | |
2.5 in. | |
10 to 90% | |
65 | |
700 to 1300hpa | |
10 sec. | |
-30°C to +50°C | |
106g | |
8.4 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction