Learn More
Dräger™ Pac™ 5500 Single-Gas Detector
Unlimited instrument life
Manufacturer:  Dräger™ 4543961
 View more versions of this product
Catalog No. 17-985-681
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
-20° to +120°F
Audible, visual, vibrating
1 in.
2.5 in.
10 to 90%
65
700 to 1300hpa
15 sec.
3.8 oz.
3.3 in.
-30° to +50°C
8 hr.
2.5cm
6.4cm
Alligator clip
0 to 100
0.1
Hydrogen Sulfide
106g
8.4cm