Dräger™ Pac™ III Sensors

Provide fast and accurate responses to hazardous gas levels

Manufacturer: Dräger™ 6809110

 View more versions of this product

Catalog No. 19-815-829

  Check Availability
Add to Cart

    Specifications

    Warranty 3 Years
    Measurement Range 0 to 100
    Pressure Range 700 to 1300mbar
    Sensor Types Hydrogen Sulfide (XS)
    Temperature (English) Range -4° to +122°F
    Temperature (Metric) Range -20° to +50°C
    Humidity Range 10 to 90%
    Resolution 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit