Dräger™ Short-Term Detector Tubes: Alcohol
Detector tubes
Manufacturer: Dräger™ CH29701
 View more versions of this product
Catalog No. 17-985-176
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Description Detector Tubes
    Gases Measured Alcohol
    Measurement Range 100 to 3000
    No. of Strokes 10
    Quantity 10
    Temperature (English) Range 59° to 77°F
    Humidity Range Less than20mg/L
    Measurement Time 1.5 min.
    Temperature (Metric) Range 15° to 25°C
    Tube Type Alcohol 100/a
  • Description & Specifications
Related Products
Product Content Feedback