Learn More
Dräger™ Short-Term Detector Tubes: Benzene
Detector tubes
Manufacturer:  Dräger™ 6728561
 View more versions of this product
Catalog No. 17-985-186
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Benzene
0.5 to 10
40 to 2
50° to 104°F
Benzene 0.5/a
3 to 15mg/L
15 min.
10
10° to 40°C