Dräger™ Short-Term Detector Tubes: Benzene
Detector tubes
Manufacturer: Dräger™ 8101841
 View more versions of this product
Catalog No. 17-988-416
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Gases Measured Benzene
    Humidity Range 1 to 30mg/L
    Measurement Range 0.5 to 10
    Measurement Time 20 min.
    No. of Strokes 20
    Quantity 5
    Temperature (English) Range 41° to 104°F
    Temperature (Metric) Range 5° to 40°C
    Tube Type Benzene 0.5/c
  • Description & Specifications
Related Products
Product Content Feedback