Dräger™ Short-Term Detector Tubes: Alcohol
Detector tubes
Manufacturer: Dräger™ 8101631
 View more versions of this product
Catalog No. 18-565
  Check Availability
Add to Cart
    Specifications
    Description Detector Tubes
    Gases Measured Alcohol
    Humidity Range 15mg/L
    Measurement Range 25 to 5000 (Methanol), 50 to 4000 (i-Propanol)
    Measurement Time 5 min.
    No. of Strokes 10
    Quantity 10
    Temperature (English) Range 59° to 86°F
    Temperature (Metric) Range 15° to 30°C
    Tube Type Alcohol 25/a
  • Description & Specifications
Related Products
Product Content Feedback