Dräger™ Short-Term Detector Tubes - Nitrogen Fumes
Manufacturer: Dräger™ CH29401
 View more versions of this product
Catalog No. 17-985-272
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Humidity Range 40mg/L
    Measurement Range 0.5 to 10ppm
    Temperature (English) Range 32° to 104°F
    Gases Measured Nitrous fumes 0.5/a
    Measurement Time 40 sec.
    No. of Strokes 5
    Quantity 10
    Temperature (Metric) Range 0° to 40°C
  • Description & Specifications
Related Products
Product Content Feedback