Learn More
Dräger™ Short-Term Detector Tubes - Nitrogen Fumes
Manufacturer:  Dräger™ CH27701
 View more versions of this product
Catalog No. 17-985-275
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Nitrous fumes 100/c
100 to 5000ppm
5/1+4
50° to 86°F
30mg/L
1.5 min./18 sec.
10
10° to 30°C