Learn More
Dräger™ Short-Term Detector Tubes - Perchloroethylene
Manufacturer:  Dräger™ CH30701
 View more versions of this product
Catalog No. 17-985-284
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Perchloroethylene 10/b
10 to 500ppm
3
59° to 104°F
5 to 12mg/L
40 sec.
10
15° to 40°C