Learn More
Dräger™ Short-Term Detector Tubes - Phosphine
Manufacturer:  Dräger™ CH31101
 View more versions of this product
Catalog No. 17-985-291
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Short-Term Gas Detector Tubes
40mg/L
6 min.
10
0° to 50°C
Phosphine 0.1/a
0.1 to 4ppm
10
32° to 122°F