Learn More
Dräger™ X-act™ 5000 Automatic Tube Pump
Designed for measurements with Draeger short-term tubes and for sampling tubes and systems
Specifications
For Use With (Application) For sampling of toxic gases and vapors in workplace
For Use With (Equipment) Designed for measurements with Draeger short-term tubes and for sampling tubes and systems
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Includes Price Quantity & Availability  
19-453-020
View Documents
Dräger™
4523500
X-act™ 5000 Automatic Tube Pump Tube pump, shoulder strap This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-453-021
View Documents
Dräger™
4523617
X-act™ 5000 Automatic Tube Pump Kit Tube Pump, Bar code reader, shoulder strap, NiMH battery, power supply This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Simple operation and reduced measurement time
  • Swiping bar-code loads measurement data
  • Automatic adjustment of flow for technical gases
  • No external flow meter needed
  • Connection with tube swivel
  • With segment and matrix display, barcode reader and flashing lights
X-act 5000 Automatic Tube Pump Kit:
  • Includes pump with shoulder strap and rechargeable NiMH battery
  • With power supply
Specifications
Specifications
For sampling of toxic gases and vapors in workplace
Designed for measurements with Draeger short-term tubes and for sampling tubes and systems