Learn More
Dräger™ X-am™ 5000 Multigas Detector
Super-small one-to-five gas detector
Manufacturer:  Dräger™ 4543748
 View more versions of this product
Catalog No. 17-985-615
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Audible, visual
NiMH
12.9cm
1.2 in.
No sensors
-20° to +50°C
220g
4.7cm
12 hr.
5.1 in.
67
3.1cm
-4° to +122°F
7.68 oz.
1.9 in.
1.2 x 1.9 x 5.1 in. (3.1 x 4.7 x 12.9cm)