Dräger™ X-plore™ Multi-Gas+ Cartridges
For protection against: Organic Vapors, Chloride, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Flouride, Formaldehyde, Ammonia, Methylamine, Hydrogen Sulfide
Manufacturer: Dräger™ 6738351
 View more versions of this product
Catalog No. 17-985-671
  Check Availability
Add to Cart
    Specifications
    Quantity Each
    For Use With (Equipment) N95 and r95 prefilter pads and low profile p100 filter of x-plore™ 3300/3500/5500 twin-filter respirator,N95 and r95 prefilter pads and low profile p100 filter of x-plore™ 3300/3500/5500 twin-filter respirator
    Product Type Cartridge
    For Use With (Application) For use under visors high level of user safety
    Certifications NIOSH
    Cartridge Type Multi-gas Cartridge
  • Description & Specifications
Related Products
Product Content Feedback