Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR3/TNFRSF25 Mouse anti-Human, Mouse, Alexa Fluor 405, Clone: 59204, R&D Systems™

Mouse MonoclonalAntibody

Manufacturer:  R&D Systems FAB943V100UG

Catalog No. FAB943V100


Add to cart
This item is not returnable. View return policy

Description

Description

DR3/TNFRSF25 Monoclonal antibody specifically detects DR3/TNFRSF25 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

DR3/TNFRSF25
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q93038
TNFRSF25
Mouse myeloma cell line NS0-derived recombinant human DR3 isoform 1 Gln25-Phe201 Accession # Q93038
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human DR3/TNFRSF25 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse (rm) DR3/TNFRSF25 and no cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR6, rhBAFF R, rhFas, rhGITR, rhLTR beta , rhNGF R, rhOPG, rmOX40, rhRANK, rhTAJ, rhTNF RI, rhTNF RII, or rhHVEM.
Flow Cytometry
59204
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), LARD, Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, TNFRSF25, TRAMP, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1, WSL-1
Mouse
IgG1
RUO
Primary
8718
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)