Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR6 (CD358) Rat anti-Mouse, Clone: 6B6, MilliporeSigma™

Rat Monoclonal Antibody

$145.80 - $373.00

Specifications

Antigen DR6 (CD358)
Clone 6B6
Concentration 0.5 mg/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC1594100
View Documents
MilliporeSigma
MABC1594100UL
100 μL
Each for $373.00
Add to cart
 
MABC159425
View Documents
MilliporeSigma
MABC159425UL
25 μL
Each for $145.80
Add to cart
 
Description

Description

DR6 (CD358) Monoclonal antibody specifically detects DR6 (CD358) in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

DR6 (CD358)
0.5 mg/mL
Unconjugated
Rat
Mouse
Recombinant fragment corresponding to full-length ectodomain region of mouse DR6 with an N‐terminal CD5 signal peptide and a C‐terminal 2XStrepII tag.
Monoclonal
Protein G purified
6B6
Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2b
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit