Learn More
Thermo Scientific UltiMate™ 3000 System Drain Kit
Supplier: Thermo Scientific 6040.0005
Drain kit for use with Ultimate 3000 systems. Keep your system free of water condensation with the Thermo Scientific™ System Drain Kit for Dionex™ UltiMate™ 3000 Systems. The kit includes all components required for system drainage and detailed installation instructions.