Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRAM Polyclonal Antibody
DRAM Polyclonal Antibody
Supplier: Thermo Scientific PA569473
Description
DRAM Polyclonal Antibody for Western Blot

Specifications
DRAM | |
Polyclonal | |
Unconjugated | |
DRAM1 | |
1200002N14Rik; Damage-regulated autophagy modulator; DNA damage regulated autophagy modulator 1; DNA damage-regulated autophagy modulator protein 1; DNA-damage regulated autophagy modulator 1; DRAM; Dram1; FLJ11259; hypothetical protein FLJ11259; zgc:101811 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
71712 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9DC58 | |
DRAM1 | |
synthetic peptide directed towards the middle region of human DRAM (aa 1-50). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction