Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ DRAQ5™

$209.00 - $450.50

Specifications

For Use With (Application) Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy
Storage Requirements Store at 4°C in the dark. Do not freeze.
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB016658
View Documents
Novus Biologicals™
NBP28112550UL
50 µL
Each for $209.00
Add to cart
 
NB016657
View Documents
Novus Biologicals™
NBP281125200UL
200 µL
Each for $450.50
Add to cart
 
NB016665
View Documents
Novus Biologicals™
NBP281129
500 µL
Each for $261.50
Add to cart
 
Description

Description

Specifications

Specifications

Flow Cytometry, Immunocytochemistry/Immunofluorescence, Live Imaging Microscopy
Store at 4°C in the dark. Do not freeze.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit