Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DRD3 Recombinant Rabbit Monoclonal Antibody (5B10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538398
Description
This antibody has been tested in direct-ELISA.
This gene encodes the D3 subtype of the five (D1-D5) dopamine receptors. The activity of the D3 subtype receptor is mediated by G proteins which inhibit adenylyl cyclase. This receptor is localized to the limbic areas of the brain, which are associated with cognitive, emotional, and endocrine functions. Genetic variation in this gene may be associated with susceptibility to hereditary essential tremor 1. Alternative splicing of this gene results in transcript variants encoding different isoforms, although some variants may be subject to nonsense-mediated decay (NMD).
Specifications
DRD3 | |
Recombinant Monoclonal | |
0.33 mg/mL | |
PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 | |
P19020, P35462 | |
DRD3 | |
A synthesized peptide derived from human Dopamine Receptor D3 (aa 19-33). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Western Blot | |
5B10 | |
Unconjugated | |
DRD3 | |
D(3) dopamine receptor; D3 receptor; D3DR; D3nf; D3R; Dopamine 3 receptor; Dopamine D3 receptor; dopamine receptor 3; dopamine receptor D3; dopaminergic receptor D3; Drd3; essential tremor 1; ETM1; FET1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1814, 29238 | |
-20°C or -80°C if preferred | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction