Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DRD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533485
Description
Percent identity with other species by BLAST analysis: Human (100%) Gorilla (94%) Gibbon, Monkey (88%) Marmoset (81%).
The D1 subtype is the most abundant dopamine receptor in the central nervous system. This G-protein coupled receptor stimulates adenylyl cyclase and activates cyclic AMP-dependent protein kinases. D1 receptors regulate neuronal growth and development, mediate some behavioral responses, and modulate dopamine receptor D2-mediated events. Alternate transcription initiation sites result in two transcript variants of this gene.
Specifications
DRD5 | |
Polyclonal | |
Unconjugated | |
DRD5 | |
D(1B) dopamine receptor; D(5) dopamine receptor; D1B; D1b receptor; D1beta dopamine receptor; D5R; DBDR; Dopamine 5 receptor; dopamine D5 receptor; dopamine receptor 5; dopamine receptor D1B; dopamine receptor D5; DRD 1B; DRD 5; Drd1b; DRD1L2; Drd5; Drd-5; Gpcr1; Hd5dr; MGC10601 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1816, 25195 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P21918, P25115 | |
DRD5 | |
Synthetic 16 amino acid peptide from 3rd extracellular domain of human DRD5 / Dopamine Receptor D5. | |
50 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction