Learn More
Thermo Scientific™ DreamTaq Green DNA Polymerase Promotion
Ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase
Manufacturer:  Thermo Scientific™ EP0713
 View more versions of this product
Catalog No. FEREP0713PM
Add to cart
Specifications
Specifications
Routine PCR amplification of DNA fragments up to 6kb; RT-PCR; genotyping; generation of PCR products for TA cloning
10 x 1.25mL of 10X DreamTaq Green Buffer
5 x 500U