Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Promotion
Contains green buffer that includes density reagent and two tracking dyes for direct loading of PCR products on gels
Manufacturer:  Thermo Scientific™ K1081
 View more versions of this product
Catalog No. FERK1081PM
Add to cart
Description
Description
  • DreamTaq DNA Pol,
  • dNTPs,
  • Green buffer, density reagent and dyes for direct loading,
  • supplied with Water,
  • Nuclease free
Specifications
Specifications
2X
Routine PCR amplification of DNA fragments up to 6kb from genomic DNA and up to 20kb from viral DNA; RT-PCR; Genotyping; Generation of PCR products for TA cloning
200
4 x 1.25mL