Learn More
Thermo Scientific™ DreamTaq™ Hot Start Green DNA Polymerase PROMO
Thermo Scientific DreamTaq Hot Start DNA Polymerase is product of choice when you need consistently robust amplification with minimal optimization steps.
Manufacturer:  Thermo Scientific™ EP1713
 View more versions of this product
Catalog No. FEREP1713PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.
Better reaction outcomes:
  • Higher yield of target amplicons from low template amounts.
  • Increased reaction specificity due to reliable hot-start technology.
  • Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
  • Room temperature reaction set-up.
  • Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.
  • Direct loading of PCR products on a gel with DreamTaq Green buffer.
Specifications
Specifications
Routine PCR, Hot Start PCR, Genotyping, colony PCR, RT-PCR
−20°C
3′-A
Separate components
DreamTaq Hot Start DNA Polymerase; 10X DreamTaq Green Buffer
2500 U