Learn More
Thermo Scientific™ DreamTaq™ Hot Start PCR Master Mix PROMO
Thermo Scientific DreamTaq Hot Start DNA Polymerase is product of choice when you need consistently robust amplification with minimal optimization steps.
Manufacturer:  Thermo Scientific™ K9011
 View more versions of this product
Catalog No. FERK9011PM
Add to cart
Description
Description
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.
Better reaction outcomes:
  • Higher yield of target amplicons from low template amounts.
  • Increased reaction specificity due to reliable hot-start technology.
  • Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
  • Room temperature reaction set-up.
  • Minimized optimization of Mg2™ concentration and of primer annealing temperatures due to optimized reaction buffer.
  • Simplified workflow and minimized pipetting steps with 2X master mix format.
Amplicon size (complex DNA): 6 kb
Fidelity (vs. Taq): 1X
Specifications
Specifications
Routine PCR, Hot Start PCR, Genotyping, colony PCR, RT-PCR
−20°C
3′-A
2X Master Mix
DreamTaq Hot Start PCR Master Mix, 2X; water, nuclease-free
200 rxns