Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DreamTaq™ Hot Start PCR Master Mix PROMO

Thermo Scientific DreamTaq Hot Start DNA Polymerase is product of choice when you need consistently robust amplification with minimal optimization steps.

Specifications

For Use With (Application) Routine PCR, Hot Start PCR, Genotyping, colony PCR, RT-PCR
Format 2X Master Mix
Includes DreamTaq Hot Start PCR Master Mix, 2X; water, nuclease-free
Overhang 3′-A
Storage Requirements −20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FERK9011PM
View Documents
Thermo Scientific™
K9011
200 rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
FERK9012PM
View Documents
Thermo Scientific™
K9012
1000 rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.

Better reaction outcomes:
  • Higher yield of target amplicons from low template amounts.
  • Increased reaction specificity due to reliable hot-start technology.
  • Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
  • Room temperature reaction set-up.
  • Minimized optimization of Mg2™ concentration and of primer annealing temperatures due to optimized reaction buffer.
  • Simplified workflow and minimized pipetting steps with 2X master mix format.
Amplicon size (complex DNA): 6 kb
Fidelity (vs. Taq): 1X
Specifications

Specifications

Routine PCR, Hot Start PCR, Genotyping, colony PCR, RT-PCR
DreamTaq Hot Start PCR Master Mix, 2X; water, nuclease-free
−20°C
2X Master Mix
3′-A
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit