Learn More
Thermo Scientific™ DreamTaq PCR Master Mix (2X) Promotion
Ensures higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase
Manufacturer:  Thermo Scientific™ K1071
 View more versions of this product
Catalog No. FERK1071PM
Add to cart
Description
Description
  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
Specifications
Specifications
1 X
200
Store at -20°C
4 x 1.25mL
Routine PCR amplification of DNA fragments up to 6 kb, High throughput PCR, Genotyping
Standard
DreamTaq DNA Polymerase, 2X DreamTaq buffer, dNTPs, and 4 mM MgCl2; 4 x 1.25 mL Nuclease-free water