Learn More
Drebrin 1 Mouse anti-Human, Alexa Fluor™ 750, Clone: DBN-N-03, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222344AF750
Catalog No. NP222344A70
Add to cart
Description
Description
Drebrin 1 Monoclonal antibody specifically detects Drebrin 1 in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications
Specifications
Drebrin 1
DBN-N-03
Western Blot, Flow Cytometry
D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal Drebrin Antibody (DBN-N-03) recognizes drebrin, an approximately 100-125 kDa regulator of actin cytoskeleton.
Flow Cytometry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
DBN1
Bacterially expressed N-terminal fragment of recombinant human Drebrin Antibody (DBN-N-03)
0.1 mL
Primary
1627
Human